
Isoquinoline-5-amine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1125-60-6
:
Formula:C₉H₈N₂
MDL Number:MFCD00006907
:144.18 g/mol
EINECS:214-408-3
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:127.0 to 132.0 deg-C
Specification Test Results
Frequently Bought Together