

Chlorodimethyl(pentafluorophenyl)silane (flophemesyl chloride) ≥95.0% (by GC) pentafluorophenyldimethylsilylating agent
Supplier: TCI
Certificates
About this item
CAS Number:20082-71-7
:
Formula:C₈H₆ClF₅Si
MDL Number:MFCD00000500
:260.67 g/mol
Boiling Pt:47°C
EINECS:243-506-9
UN:2985
ADR:3,II
:GHS02
:GHS05
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together