Order Entry
Netherlands
ContactUsLinkComponent
1-Aza-18-crown-6 (1,4,7,10,13-pentaoxa-16-azacyclooctadecane) ≥98.0% (by GC)
Irritant
1-Aza-18-crown-6 (1,4,7,10,13-pentaoxa-16-azacyclooctadecane) ≥98.0% (by GC)
Supplier:  TCI
Certificates
undefined
Irritant
1-Aza-18-crown-6 (1,4,7,10,13-pentaoxa-16-azacyclooctadecane) ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystaline
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 97.0 %
  • Melting point:
    50.0 to 55.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together