Order Entry
Netherlands
ContactUsLinkComponent
Tetrahydrofuran ≥99.0% (by GC) stabilised for determination of vinyl chloride monomer
Flammable
Irritant
Health
Tetrahydrofuran ≥99.0% (by GC) stabilised for determination of vinyl chloride monomer
Supplier:  TCI
Certificates
undefined
Flammable
Irritant
Health
Tetrahydrofuran ≥99.0% (by GC) stabilised for determination of vinyl chloride monomer
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.0 %
  • Suitability for VCM-analysis:
    to pass test
  • Peroxide:
    max. 0.005 %

Specification Test Results

Related Information

Frequently Bought Together