


Divinylbenzene (mixture of isomers) >50.0% (by GC) (contains Ethylvinylbenzene, Diethylbenzene) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:1321-74-0
:
Formula:C₁₀H₁₀
MDL Number:MFCD01778656
:130.19 g/mol
Flash Pt:68°C
Storage Temperature:Refrigerator
EINECS:215-325-5
UN:3082
ADR:9,III
:Stabilised with 4-tert-butylpyrocatechol (TBC)
:GHS07
:GHS08
:GHS09
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 50.0 % (total of isomers)
Specification Test Results
Frequently Bought Together