Saccharin ≥99.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:81-07-2
:
Formula:C₇H₅NO₃S
MDL Number:MFCD00005866
:183.19 g/mol
Merck Index:14,08311
EINECS:201-321-0
Specification Test Results
- Appearance:White powder to crystal
- Purity (HPLC):min. 99.0 area%
- Purity (Neutralization titration):min. 99.0 %
- Melting point:226.0 to 233.0 deg-C
Specification Test Results
Frequently Bought Together