
Methyltrioctylammonium bis(trifluoromethylsulphonyl)imide ≥98.0% (by total nitrogen basis)
Catalog # TCIAM1660-5G
Supplier: TCI
CAS Number:
Specifications
- Verpakking:Glass bottle
- VPE:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange clear liquid
- Purity (with Total Nitrogen):min. 98.0 %
Specification Test Results
About this item
:
:648.86 g/mol
:GHS07
CAS Number:375395-33-8
Formula:C₂₇H₅₄F₆N₂O₄S₂
MDL Number:MFCD08275364