Order Entry
Netherlands
ContactUsLinkComponent
Sodium benzoate ≥98.0% (by HPLC, titration analysis) for biochemical research
Sodium benzoate ≥98.0% (by HPLC, titration analysis) for biochemical research
Supplier:  TCI
Certificates
undefined
Sodium benzoate ≥98.0% (by HPLC, titration analysis) for biochemical research
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Ion exchange titration):
    min. 98.0 %
  • Protein impurity test (SDS-PAGE):
    to pass test

Specification Test Results

Related Information

Frequently Bought Together