

3,4-Epoxy-1-butene ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:930-22-3
:
Formula:C₄H₆O
MDL Number:MFCD00005149
:70.09 g/mol
Boiling Pt:67°C
Melting Pt:–135°C
Density:0,872 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:213-210-4
UN:1993
ADR:3,II
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid to slightly cloudy liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together