Order Entry
Netherlands
ContactUsLinkComponent
1,3-Diiodopropane ≥97.0% (by GC) stabilised
Irritant
1,3-Diiodopropane ≥97.0% (by GC) stabilised
Catalog # TCIAD0611-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
1,3-Diiodopropane ≥97.0% (by GC) stabilised
Catalog # TCIAD0611-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Red to Green clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :295.89 g/mol
Boiling Pt:111°C
Melting Pt:–20 °C
Density:2,59617 g/cm³ (4 °C)
 :Stabilised with copper chip
EINECS:210-993-4
 :GHS07
CAS Number:627-31-6
Formula:C₃H₆I₂
MDL Number:MFCD00001097