
2-Adamantylamine hydrochloride ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:10523-68-9
:
Formula:C₁₀H₁₈ClN
MDL Number:MFCD00074743
:187.71 g/mol
Melting Pt:300 °C
EINECS:234-074-2
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Argentmetric Titration):min. 99.0 %
- Solubility in Water:very faint turbidity
Specification Test Results
Frequently Bought Together