

1,3-Dimethylcyclohexane cis- and trans-mixture ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:591-21-9
:
Formula:C₈H₁₆
MDL Number:MFCD00064173
:112.22 g/mol
Boiling Pt:120 °C (1013 hPa)
Density:0,7672 g/cm³ (20 °C)
Flash Pt:6°C
EINECS:209-707-0
UN:2263
ADR:3,II
:GHS02
:GHS08
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 % (total of isomers)
Specification Test Results
Frequently Bought Together