Order Entry
Netherlands
ContactUsLinkComponent
2,6-Dimethylcyclohexanone mixture of isomers ≥98.0% (by GC)
Flammable
2,6-Dimethylcyclohexanone mixture of isomers ≥98.0% (by GC)
Supplier:  TCI
Certificates
undefined
Flammable
2,6-Dimethylcyclohexanone mixture of isomers ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 98.0 % (total of isomers)

Specification Test Results

Frequently Bought Together