Order Entry
Netherlands
ContactUsLinkComponent
 
 
 
 
 
1,3-Dichloropropene cis- and trans-mixture ≥92.0% (by GC)
 :  TCI
 
undefined
 
 
 
 
1,3-Dichloropropene cis- and trans-mixture ≥92.0% (by GC)
 :  TCI

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 92.0 % (total of isomers)

 

Frequently Bought Together