
ω-Chloro-3,4-dihydroxyacetophenone ≥98.0% (by HPLC, titration analysis)
TCIAC0156-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Appearance:Light yellow to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Solubility in Methanol:almost transparency
:
:186.59
:202-754-8
:GHS07
:99-40-1
:C₈H₇ClO₃
:MFCD00002200