
1,4-Bis(5-phenyloxazol-2-yl)benzene ≥98.0% (by HPLC, total nitrogen) for scintillation spectrometry
Supplier: TCI
Certificates
About this item
CAS Number:1806-34-4
:
Formula:C₂₄H₁₆N₂O₂
MDL Number:MFCD00005309
:364.4 g/mol
Melting Pt:252 °C
EINECS:217-304-6
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Light yellow to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:242.0 to 248.0 deg-C
- Solubility in hot Toluene:almost transparency
Specification Test Results
Frequently Bought Together