

Tetranitro blue tetrazolium ≥96.0% (by HPLC, titration analysis) for biochemical research
Catalog # TCIAT0250-100MG
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (HPLC):min. 96.0 area%
- Purity (Argentmetric Titration):min. 96.0 %
- Alkaline Phosphatase detection test:to pass test
Specification Test Results
About this item
:
:907.64 g/mol
Melting Pt:165 °C
EINECS:214-665-1
UN:1325
ADR:4.1,III
:GHS02
:GHS07
CAS Number:1184-43-6
:42798-98-1
Formula:C₄₀H₂₈Cl₂N₁₂O₁₀
MDL Number:MFCD00036338