(4-Bromophenoxy)acetic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1878-91-7
:
Formula:(C₆H₄Br)OCH₂COOH
MDL Number:MFCD00093070
:231.05 g/mol
Boiling Pt:322 °C (1013 hPa)
Melting Pt:161.4…161.8 °C
EINECS:217-530-5
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together