
2-Picoline-N-oxide ≥99.0% (by GC)
: TCI
:931-19-1
:
:C₆H₇NO
:MFCD00006199
:109.13 g/mol
:259 °C (1013 hPa)
:0 °C
:213-230-3
:GHS07
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 99.0 %
- Solubility in Water:almost transparency
Frequently Bought Together