

tert-Butylchlorodimethylsilane ≥98.0% (by GC) tert-butyl-dimethyl-silylating agent
Supplier: TCI
Certificates
About this item
CAS Number:18162-48-6
:
Formula:C₆H₁₅ClSi
MDL Number:MFCD00000501
:150.72 g/mol
Boiling Pt:125°C
Melting Pt:87…90 °C
Density:0,81 g/cm³ (20 °C)
Flash Pt:22 °C
EINECS:242-042-4
UN:2925
ADR:4.1,II
:GHS02
:GHS05
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:88.0 to 92.0 deg-C
Specification Test Results
Related Information
Frequently Bought Together