


N,N-Dimethylformamide dimethyl acetal ≥98.0% (by GC) for esterification
Catalog # TCIAD2071-25ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Specific gravity (20/20):0.8940 to 0.8980
- Refractive index n20/D:1.3950 to 1.3980
- High boiling impurities:No impurity higher than Isopropyl myristate (50 ppm) after 5min + hold up time.
Specification Test Results
About this item
:
:119.16 g/mol
Boiling Pt:104°C
Density:0,893 g/cm³ (20 °C)
Flash Pt:5°C
EINECS:225-063-3
UN:1993
ADR:3,II
:GHS02
:GHS07
:GHS08
CAS Number:4637-24-5
Formula:C₅H₁₃NO₂
MDL Number:MFCD00008482