

Ethyl formate ≥98.0% (by GC) for spectrophotometry
Catalog # TCIAF0085-100ML
Leverancier: TCI
CAS-nummer:
Specificaties
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specificaties
Specificatie testresultaten
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Absorbance (270nm):max. 1.000
- Absorbance (280nm):max. 0.020
- Absorbance (290nm):max. 0.010
Specificatie testresultaten
Over dit artikel
:
:74.08 g/mol
Merck index:14,03807
Kokend Pt:54°C
Smelten Pt:-80°C
Smelten Pt:–80°C
Dichtheid:0,9208 g/cm³ (20 °C)
Flits Pt:-20°C
Flits Pt:–20°C
Temperatuur bij opslag:Ambient
EINECS:203-721-0
UN:1190
ADR:3,II
:GHS02
:GHS07
CAS-nummer:109-94-4
Formule:C₃H₆O₂
MDL-nummer:MFCD00003294