Order Entry
Netherlands
ContactUsLinkComponent
Hydrazinium hydroxide
Corrosive
Toxic
Health
Environment
Hydrazinium hydroxide
Supplier:  TCI
Certificates
undefined
Corrosive
Toxic
Health
Environment
Hydrazinium hydroxide
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless clear liquid
  • Concentration (Potassium iodate Method):
    79.0 to 82.0 w/w%
  • Solubility in Water:
    transparency

Specification Test Results

Related Information

Frequently Bought Together