Order Entry
Netherlands
ContactUsLinkComponent
 
 
 
2,6-Dimethyl-3-heptene cis- and trans-mixture ≥98.0% (by GC)
  TCIAD1262-0.1ML
 :  TCI
 :  
undefined
 
 
2,6-Dimethyl-3-heptene cis- and trans-mixture ≥98.0% (by GC)
  TCIAD1262-0.1ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    0,1 mL
  • Seq:
    2010

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %

 

 

 :
 :126.24 g/mol
 :134°C
 :0,7311 g/cm³ (20 °C)
 :15°C
 :220-362-5
 :3295
 :3,II
 :GHS02
 :GHS08
 :2738-18-3
 :C₉H₁₈
 :MFCD00026523