2-Heptene cis- and trans-mixture ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:592-77-8
:
Formula:C₇H₁₄
MDL Number:MFCD00009506
:98.19 g/mol
Boiling Pt:98,5 °C (1013 hPa)
Density:0,7035 g/cm³ (20 °C)
Flash Pt:–1°C
EINECS:209-768-3
UN:2278
ADR:3,II
:GHS02
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 % (total of isomer)
Specification Test Results
Frequently Bought Together