1,2,4,5-Tetramethyl-3,6-dinitrobenzene ≥97.0% (by GC)
TCIAD1661-25G
: TCI
:
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
- Purity (GC):min. 97.0 %
- Melting point:210.0 to 214.0 deg-C
: 1,4-Dinitrodurene, Dinitrodurene
:
:224.22 g/mol
:226-766-8
:5465-13-4
:C₁₀H₁₂N₂O₄
:MFCD00007164
:363