Order Entry
Netherlands
ContactUsLinkComponent
 
 
Decafluoro-4,6-bis(trifluoromethyl)cyclohexane ≥80.0% (by GC)
  TCIAP1420-25G
 :  TCI
 :  
undefined
 
Decafluoro-4,6-bis(trifluoromethyl)cyclohexane ≥80.0% (by GC)
  TCIAP1420-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 80.0 % (total of isomer)

 

 

 :
 :400.06 g/mol
 :206-386-9
 :GHS07
 :335-27-3
 :C₈F₁₆
 :MFCD00001469