
1-Chloro-3,3-dimethylbutanone ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:13547-70-1
:
Formula:C₆H₁₁ClO
MDL Number:MFCD00035688
:134.61 g/mol
Boiling Pt:173°C
Melting Pt:–12.6 °C
Density:1,1273 g/cm³ (20 °C)
Flash Pt:67°C
Storage Temperature:Refrigerator
EINECS:236-920-6
ADR:6.1,I
:GHS07
Specification Test Results
- Appearance:Colorless to Yellow to Green clear liquid
- Purity (GC):min. 97.0 %
Specification Test Results
Frequently Bought Together