Order Entry
Netherlands
ContactUsLinkComponent
 
 
1,2,3-Trichloropropene cis- and trans-mixture ≥95.0% (by GC)
 :  TCI
 
undefined
 
1,2,3-Trichloropropene cis- and trans-mixture ≥95.0% (by GC)
 :  TCI

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 95.0 % (total of isomer)

 

Frequently Bought Together