Order Entry
Netherlands
ContactUsLinkComponent
 
 
 
Benzenesulphonic acid, anhydrous ≥98.0% (by titrimetric analysis)
  TCIAB3408-5G
 :  TCI
 :  
undefined
 
 
Benzenesulphonic acid, anhydrous ≥98.0% (by titrimetric analysis)
  TCIAB3408-5G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Neutralization titration):
    min. 98.0 %
  • Melting point:
    58.0 to 64.0 deg-C
  • Diphenyl sulfone:
    max. 2.0 area%

 

 

 :
 :158.18 g/mol
 :14,01070
 :319 °C (1013 hPa)
 :50 °C
 :1,32 g/cm³ (47 °C)
 :≥ 100 °C
 :Ambient
 :202-638-7
 :2585
 :8,III
 :GHS05
 :GHS07
 :98-11-3
 :C₆H₆O₃S
 :MFCD00011689