
7-Amino-4-methylcoumarin ≥98.0% (by HPLC, titration analysis) for HPLC labeling
Supplier: TCI
Certificates
About this item
CAS Number:26093-31-2
:
Formula:C₁₀H₉NO₂
MDL Number:MFCD00006868
:175.19 g/mol
Boiling Pt:352 °C (1013 hPa)
Melting Pt:226 °C
Density:1,377 g/cm³ (20 °C)
EINECS:247-454-8
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown to Dark green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:224.0 to 230.0 deg-C
Specification Test Results
Frequently Bought Together