(S)-(-)-2-Methyl-CBS-oxazaborolidine ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:112022-81-8
:
Formula:C₁₈H₂₀BNO
MDL Number:MFCD00078439
:277.17 g/mol
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 95.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Specific rotation [a]20/D:-66.0 to -73.0 deg (C=1, MeOH)
Specification Test Results
Frequently Bought Together