(R)-(+)-2-Methyl-CBS-oxazaborolidine ≥97.0% (by titrimetric analysis)
: TCI
:112022-83-0
:
:C₁₈H₂₀BNO
:MFCD00078440
:277.17 g/mol
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 97.0 %
- Specific rotation [a]20/D:+66.0 to +73.0 deg (C=1, MeOH)
Frequently Bought Together