


(S)-(-)-2-Methyl-CBS-oxazaborolidine 1 M in toluene
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:112022-81-8
:
Formula:C₁₈H₂₀BNO
MDL Number:MFCD00078439
:277.17 g/mol
Density:0.925
Flash Pt:4 °C (39 °F)
UN:1294
ADR:3,II
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together