Order Entry
Netherlands
ContactUsLinkComponent
Benzo[a]pyrene-[D12] 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Benzo[a]pyrene-[D12] 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERLA20635100AL
Supplier:  EHRENSTORFER
CAS Number:  
Benzo[a]pyrene-[D12] 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERLA20635100AL
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    63466-71-7
  • Description:
    Benzo[a]pyrene-[D12] 10 µg/ml in acetonitrile
  • Formula:
    C₂₀D₁₂
  • Description:
    Benzo[a]pyrene-[D12]

Specifications

About this item