Diethyl cyclohexane-1,1-dicarboxylate ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1139-13-5
:
Formula:C₁₂H₂₀O₄
MDL Number:MFCD02093457
:228.29 g/mol
Boiling Pt:137°C
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together