Sodium decanoate ≥99.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1002-62-6
:
Formula:C₁₀H₁₉NaO₂
MDL Number:MFCD00066453
:194.2464 g/mol
EINECS:213-688-4
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Nonaqueous Titration):min. 99.0% (calcd.on dried.substance)
- Solubility in Water:very faint turbidity
- Drying loss:max. 3.0 %
Specification Test Results
Frequently Bought Together