Order Entry
Netherlands
ContactUsLinkComponent
Isopentyl decanoate ≥97.0% (by GC)
Isopentyl decanoate ≥97.0% (by GC)
Catalog # TCIAD0019-25ML
Supplier:  TCI
CAS Number:  
undefined
Isopentyl decanoate ≥97.0% (by GC)
Catalog # TCIAD0019-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :242.3975 g/mol
Boiling Pt:124°C
Flash Pt:148°C
EINECS:218-982-6
CAS Number:2306-91-4
Formula:C₁₅H₃₀O₂
MDL Number:MFCD00053822