Order Entry
Netherlands
ContactUsLinkComponent
Butamben (butyl-4-aminobenzoate) ≥99.0% (by GC, titration analysis)
Butamben (butyl-4-aminobenzoate) ≥99.0% (by GC, titration analysis)
  TCIAA0270-25G
 :  TCI
 :  
undefined
Butamben (butyl-4-aminobenzoate) ≥99.0% (by GC, titration analysis)
  TCIAA0270-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (GC):
    min. 99.0 %
  • Purity (Nonaqueous Titration):
    min. 99.0 %
  • Melting point:
    57.0 to 61.0 deg-C
  • Solubility in Methanol:
    almost transparency

 

 

 :
 :193.25 g/mol
 :14,01512
 :202-317-1
 :GHS07
 :94-25-7
 :C₁₁H₁₅NO₂
 :MFCD00017112