



Diethylbenzene (mixture of isomers) ≥70% (by GC)
Catalog # TCIAD0480-25ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 70.0 %
Specification Test Results
About this item
:
:134.22 g/mol
Boiling Pt:184°C
Melting Pt:-31°C
Melting Pt:–31°C
Flash Pt:55°C
EINECS:246-874-9
UN:2049
ADR:3,III
:GHS02
:GHS07
:GHS08
:GHS09
CAS Number:25340-17-4
Formula:C₁₀H₁₄
MDL Number:MFCD00792900