



Thionyl chloride ∼1 mol/L in dichloromethane
Catalog # TCIAT2048-500ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Concentration (Argentmetric titration):9.0 to 11.5 w/w%
Specification Test Results
About this item
:
:118.97 g/mol
Merck Index:14,09348
EINECS:231-748-8
UN:2922
ADR:8,II
:GHS05
:GHS06
:GHS08
:GHS09
CAS Number:7719-09-7
Formula:Cl₂OS
MDL Number:MFCD00011449