Order Entry
Netherlands
ContactUsLinkComponent
 
 
Stearic acid hydrazide ≥95.0% (by GC, titration analysis)
  TCIAS0222-25G
 :  TCI
 :  
undefined
 
Stearic acid hydrazide ≥95.0% (by GC, titration analysis)
  TCIAS0222-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 95.0 %
  • Purity (Nonaqueous Titration):
    min. 95.0 %
  • Melting point:
    114.0 to 118.0 deg-C
  • Solubility in hot Methanol:
    almost transparency

 

 

 :
 :298.51 g/mol
 :223-946-8
 :GHS07
 :4130-54-5
 :C₁₈H₃₈N₂O
 :MFCD00014764