
2-(Methoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
Leverancier: TCI
Certificaten
Over dit artikel
CAS-nummer:374538-03-1
:
Formule:C₈H₉BO₄
MDL-nummer:MFCD02179452
Smelten Pt:104°C
:179.97 g/mol
:GHS07
Specificatie testresultaten
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (Neutralization titration):97.0 to 111.0 %
Specificatie testresultaten
Gerelateerde informatie
Frequently Bought Together