Order Entry
Netherlands
ContactUsLinkComponent
1-Aza-15-crown-5 (1,4,7,10-tetraoxa-13-azacyclopentadecane) ≥98.0% (by titrimetric analysis)
Irritant
1-Aza-15-crown-5 (1,4,7,10-tetraoxa-13-azacyclopentadecane) ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Irritant
1-Aza-15-crown-5 (1,4,7,10-tetraoxa-13-azacyclopentadecane) ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystaline
  • Purity (GC):
    min. 96.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    41.0 to 46.0 deg-C

Specification Test Results

Related Information

Frequently Bought Together