

4,4'-Bis(dimethylamino)diphenylmethane ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:101-61-1
:
Formula:C₁₇H₂₂N₂
MDL Number:MFCD00008317
:254.38 g/mol
Merck Index:14,06176
EINECS:202-959-2
:GHS08
:GHS09
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:88.0 to 92.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
Frequently Bought Together