Order Entry
Netherlands
ContactUsLinkComponent
4-Chloromethyl-o-xylene ≥70% (by GC) (contains isomer)
Corrosive
4-Chloromethyl-o-xylene ≥70% (by GC) (contains isomer)
Catalog # TCIAD0290-25G
Supplier:  TCI
CAS Number:  
undefined
Corrosive
4-Chloromethyl-o-xylene ≥70% (by GC) (contains isomer)
Catalog # TCIAD0290-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 70.0 %

Specification Test Results

About this item

 :
Boiling Pt:117°C
EINECS:203-032-5
UN:3265
ADR:8,II
CAS Number:102-46-5