
2-(Methylthio)phenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI
Certificates
About this item
CAS Number:168618-42-6
:
Formula:C₇H₉BO₂S
MDL Number:MFCD01318165
:168.02 g/mol
:GHS07
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (Neutralization titration):97.0 to 112.0 %
Specification Test Results
Related Information
Frequently Bought Together