Order Entry
Netherlands
ContactUsLinkComponent
1,4-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
Irritant
1,4-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
1,4-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystaline
  • Purity (GC):
    min. 98.0 % (total of isomers)
  • Purity (Neutralization titration):
    min. 98.0 %
  • Solubility in Methanol:
    very faint turbidity

Specification Test Results

Related Information

Frequently Bought Together