Order Entry
Netherlands
ContactUsLinkComponent
1,3-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
Irritant
1,3-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
1,3-Cyclohexanedicarboxylic acid (mixture of cis and trans isomers) cis- and trans-mixture ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0% (total of isomer)
  • Purity (Neutralization titration):
    min. 98.0 %

Specification Test Results

Related Information

Frequently Bought Together