
1-Iodoheptane ≥98.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:4282-40-0
:
Formula:C₇H₁₅I
MDL Number:MFCD00001104
:226.1 g/mol
Boiling Pt:204 °C (1013 hPa)
Melting Pt:–48 °C
Density:1,38 g/cm³ (20 °C)
Flash Pt:78°C
:Stabilised with copper chip
EINECS:224-285-8
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to Brown clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together